Abnova
Manufacturer Code:PAB13384
Catalog # 89-116-631
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to N-terminus 14 amino acids of human SLC35D1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SLC35D1.
For Research Use Only
Protein Aliases: KIAA0260; MGC138236; solute carrier family 35 (UDP-GlcA/UDP-GalNAc transporter), member D1; solute carrier family 35 (UDP-glucuronic acid/UDP-N-acetylgalactosamine dual transporter), member D1; Solute carrier family 35 member D1; UDP-galactose transporter-related 7; UDP-galactose transporter-related protein 7; UDP-GlcA/UDP-GalNAc transporter; UDP-glucuronic acid/UDP-N-acetylgalactosamine transporter; UGTREL7
Gene Aliases: KIAA0260; SLC35D1; UGTREL7
UniProt ID: (Human) Q9NTN3
Entrez Gene ID: (Human) 23169
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.