Invitrogen
Manufacturer Code:PA542475
Catalog # PIPA542475
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SLC35C1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GDP-fucose transporter; GDP-fucose transporter 1; golgi GDP-fucose translocator; multispan transmembrane protein fuct1; solute carrier family 35 (GDP-fucose transporter), member C1; Solute carrier family 35 member C1; solute carrier family 35, member C1
Gene Aliases: CDG2C; E430007K15Rik; FUCT1; GDP-Fuc-Tr; SLC35C1
UniProt ID: (Bovine) A6QM03, (Human) Q96A29, (Mouse) Q8BLX4
Entrez Gene ID: (Rabbit) 100348541, (Bovine) 540413, (Horse) 100050209, (Pig) 397229, (Dog) 403593, (Human) 55343, (Mouse) 228368, (Rat) 311204, (Guinea pig) 100724079
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.