Invitrogen
Manufacturer Code:PA562043
Catalog # PIPA562043
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SLC35B4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: solute carrier family 35 (UDP-xylose/UDP-N-acetylglucosamine transporter), member B4; Solute carrier family 35 member B4; solute carrier family 35, member B4; UDP-xylose and UDP-N-acetylglucosamine transporter; UDP-Xylose/N-Acetylglucosamine transporter; YEA YEA4 UDP-Xylose/N-Acetylglucosamine transporter UDP-xylose and UDP-N-acetylglucosamine transporter YEA4 homolog; YEA4 homolog
Gene Aliases: PSEC0055; SLC35B4; YEA; YEA4
UniProt ID: (Human) Q969S0
Entrez Gene ID: (Human) 84912
Molecular Function: carbohydrate transporter mitochondrial carrier protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.