Invitrogen
Manufacturer Code:PA542450
Catalog # PIPA542450
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human SLC35B1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Endoplasmic reticulum nucleotide sugar transporter 1; galactose transporter; hUGTrel1; putative endoplasmic reticulum nucleotide sugar transporter; Solute carrier family 35 member B1; solute carrier family 35 member B1-like protein; solute carrier family 35, member B1; UDP-galactose translocator 2; UDP-galactose transporter related; UDP-galactose transporter-related protein 1; UGTrel1
Gene Aliases: ERNST; SLC35B1; UGALT2; UGTREL1; wu:fj99g11; zgc:92284
UniProt ID: (Bovine) Q8MII5, (Human) P78383, (Rat) Q6V7K3, (Mouse) P97858, (Zebrafish) Q66HX0
Entrez Gene ID: (Dog) 480549, (Horse) 100069752, (Rabbit) 100343677, (Bovine) 353217, (Human) 10237, (Rat) 287642, (Mouse) 110172, (Guinea pig) 100712906, (Zebrafish) 447844
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.