Invitrogen
Manufacturer Code:PA553292
Catalog # PIPA553292
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SLC35A3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Golgi UDP-GlcNAc transporter; solute carrier family 35 (UDP-N-acetylglucosamine (UDP-GlcNAc) transporter), member 3; solute carrier family 35 (UDP-N-acetylglucosamine (UDP-GlcNAc) transporter), member A3; Solute carrier family 35 member A3; UDP-N-acetylglucosamine transporter
Gene Aliases: AMRS; SLC35A3
UniProt ID: (Human) Q9Y2D2
Entrez Gene ID: (Human) 23443
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.