Invitrogen
Manufacturer Code:PA542454
Catalog # PIPA542454
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SLC35A3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Golgi UDP-GlcNAc transporter; solute carrier family 35 (UDP-N-acetylglucosamine (UDP-GlcNAc) transporter), member 3; solute carrier family 35 (UDP-N-acetylglucosamine (UDP-GlcNAc) transporter), member A3; solute carrier family 35 member 3; solute carrier family 35 member 3A; Solute carrier family 35 member A3; UDP-N-acetylglucosamine transporter
Gene Aliases: AMRS; SLC35A3
UniProt ID: (Human) Q9Y2D2, (Bovine) Q6YC49, (Dog) O77592, (Rat) Q6AXR5
Entrez Gene ID: (Human) 23443, (Horse) 100056015, (Bovine) 520918, (Dog) 442991, (Rabbit) 100348245, (Rat) 310808
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.