Invitrogen
Manufacturer Code:PA571363
Catalog # PIPA571363
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C terminal region of mouse SLC35A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: mUGT1; solute carrier family (UDP-N-acetylglucosamine transporter), member 2; solute carrier family 35 (UDP-galactose transporter) member 2; solute carrier family 35 (UDP-galactose transporter), member 2; Solute carrier family 35 member A2; UDP-Gal-Tr; UDP-galactose translocator; UDP-galactose transporter; UGALT UGAT UGT UGT1 UGT2 UGTL UDP-Gal-Tr UDP-galactose translocator UDP-galactose transporter solute carrier family 35 (UDP-galactose transporter) member 2 solute carrier family 35 member A2
Gene Aliases: AI327289; Had-1; Had1; Sfc8; Slc35a2; Ugalt; UGT; Ugt1
UniProt ID: (Mouse) Q9R0M8
Entrez Gene ID: (Mouse) 22232
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.