Invitrogen
Manufacturer Code:PA568493
Catalog # PIPA568493
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C terminal region of human SLC35A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: solute carrier family 35 (UDP-galactose transporter), member 2; solute carrier family 35 (UDP-galactose transporter), member A2; Solute carrier family 35 member A2; UDP-Gal-Tr; UDP-galactose translocator; UDP-galactose transporter; UGALT UGAT UGT UGT1 UGT2 UGTL UDP-Gal-Tr UDP-galactose translocator UDP-galactose transporter solute carrier family 35 (UDP-galactose transporter) member 2 solute carrier family 35 member A2
Gene Aliases: CDG2M; CDGX; SLC35A2; UDP-Gal-Tr; UGALT; UGAT; UGT; UGT1; UGT2; UGTL
UniProt ID: (Human) P78381
Entrez Gene ID: (Human) 7355
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.