Proteintech
Manufacturer Code:136571AP150UL
Catalog # 50-172-8913
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC35A2 Fusion Protein Ag4524 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: mUGT1; SLC35A2 UDP Gal Tr UDP galactose translocator UDP galactose transporter UGALT UGAT UGT UGT1 UGT2 UGTL; solute carrier family (UDP-N-acetylglucosamine transporter), member 2; solute carrier family 35 (UDP-galactose transporter) member 2; solute carrier family 35 (UDP-galactose transporter), member 2; solute carrier family 35 (UDP-galactose transporter), member A2; Solute carrier family 35 member A2; solute carrier family 35, member A2; UDP-Gal-Tr; UDP-galactose translocator; UDP-galactose transporter
Gene Aliases: AI327289; CDG2M; CDGX; Had-1; Had1; Sfc8; SLC35A2; UDP-Gal-Tr; UGALT; UGAT; UGT; UGT1; UGT2; UGTL
UniProt ID: (Human) P78381, (Mouse) Q9R0M8
Entrez Gene ID: (Human) 7355, (Mouse) 22232, (Rat) 100158233
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.