Proteintech
Manufacturer Code:163421AP150UL
Catalog # 50-173-0882
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC35A1 Fusion Protein Ag9524 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CMP SA Tr CMP Sia Tr CMP sialic acid transporter CMPST CST hCST inactive SLC35A1; CMP-SA-Tr; CMP-Sia-Tr; CMP-sialic acid transporter; mutated CMP-sialic acid transporter A1; solute carrier family 35 (CMP-sialic acid transporter), member 1; solute carrier family 35 (CMP-sialic acid transporter), member A1; solute carrier family 35 (UDP-galactose transporter), member 1; Solute carrier family 35 member A1
Gene Aliases: AA408150; AI314851; CDG2F; CMPST; CST; hCST; SLC35A1
UniProt ID: (Human) P78382, (Mouse) Q61420
Entrez Gene ID: (Human) 10559, (Rat) 313139, (Mouse) 24060
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.