Invitrogen
Manufacturer Code:PA595706
Catalog # PIPA595706
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 210-320 of human SLC34A3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Na(+)-dependent phosphate cotransporter 2C; Na(+)/Pi cotransporter 2C; Na+/Pi-cotransporter type IIc; naPi-2c; Sodium-dependent phosphate transport protein 2C; sodium-phosphate cotransporter IIC; Sodium-phosphate transport protein 2C; Sodium/inorganic phosphate cotransporter IIC; Sodium/phosphate cotransporter 2C; solute carrier family 34 (sodium phosphate), member 3; solute carrier family 34 (type II sodium/phosphate contransporter), member 3; solute carrier family 34 (type II sodium/phosphate cotransporter), member 3; Solute carrier family 34 member 3; type IIc Na+/Pi cotransporter; type IIc Na+/Pi-cotransporter; type IIc NaPi-cotransporter-v1
Gene Aliases: AI649385; HHRH; naPi-2c; NPT2C; Npt2c-v1; NPTIIC; SLC34A3
UniProt ID: (Human) Q8N130, (Rat) Q8K4R8, (Mouse) Q80SU6
Entrez Gene ID: (Human) 142680, (Rat) 246234, (Mouse) 142681
Molecular Function: enzyme modulator transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.