Invitrogen
Manufacturer Code:PA5100689
Catalog # PIPA5100689
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SLC34A2 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA536683 D5Ertd227e Na(+)/Pi cotransporter 2B Na(+)-dependent phosphate cotransporter 2B NAPI IIb NaPi-2b NaPi3b NAPI-3B NAPI-IIb Npt2b NPTIIb rNaPi IIb SLC34A2 Sodium/phosphate cotransporter 2B Sodium-dependent phosphate transport protein 2B Sodium-phosphate transport protein 2B solute carrier family 34 (sodium phosphate) member 2 solute carrier family 34 (type II sodium/phosphate contransporter) member 2 solute carrier family 34 (type II sodium/phosphate cotransporter) member 2 solute carrier family 34 member 2 type II sodium-dependent phosphate transporter 3b type IIb Na/Picotransporter type IIb sodium-phosphate transporter; Na(+)-dependent phosphate cotransporter 2B; Na(+)/Pi cotransporter 2B; NaPi3b; Sodium-dependent phosphate transport protein 2B; Sodium-phosphate transport protein 2B; Sodium/phosphate cotransporter 2B; solute carrier family 34 (sodium phosphate), member 2; solute carrier family 34 (type II sodium/phosphate cotransporter), member 2; Solute carrier family 34 member 2; type II sodium-dependent phosphate transporter 3b; type IIb Na/Picotransporter
Gene Aliases: AA536683; D5Ertd227e; NaPi-2b; NAPI-3B; NAPI-IIb; Npt2b; NPTIIb; SLC34A2
UniProt ID: (Human) O95436, (Mouse) Q9DBP0
Entrez Gene ID: (Human) 10568, (Mouse) 20531
Molecular Function: enzyme modulator transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.