Abnova
Manufacturer Code:PAB23198
Catalog # 89-124-898
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human SLC34A2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant SLC34A2.
For Research Use Only
Protein Aliases: FLJ90534; Na(+)-dependent phosphate cotransporter 2B; Na(+)/Pi cotransporter 2B; NAPI-3B; NAPI-IIb; NaPi3b; NPTIIb; Sodium-dependent phosphate transport protein 2B; Sodium-phosphate transport protein 2B; Sodium/phosphate cotransporter 2B; solute carrier family 34 (sodium phosphate), member 2; solute carrier family 34 (type II sodium/phosphate cotransporter), member 2; Solute carrier family 34 member 2; type II sodium-dependent phosphate transporter 3b
Gene Aliases: NAPI-3B; NAPI-IIb; NPTIIb; SLC34A2
UniProt ID: (Human) O95436
Entrez Gene ID: (Human) 10568
Molecular Function: enzyme modulator transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.