Novus Biologicals
Manufacturer Code:NBP242216APC
Catalog # NBP242216A
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 10B1.3E9 |
Immunogen | Two synthetic peptides made to an internal portions of the human SLC34A1 protein (between amino acids 25-150 and 100-200) [UniProt Q06495] |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FRTS2 Na(+)/Pi cotransporter 2A Na(+)-dependent phosphate cotransporter 2A NaPi-2a NaPi-3 NPT2NPHLOP1 NPTIIa renal sodium-dependent phosphate transporter SLC11 SLC17A2Na+-phosphate cotransporter type II sodium/phosphate co-transporter Sodium/phosphate cotransporter 2A sodium-dependent phosphate transport protein 2A Sodium-phosphate transport protein 2A solute carrier family 17 (sodium phosphate) member 2 solute carrier family 34 (sodium phosphate) member 1 Solute carrier family 34 member 1; Na(+)-dependent phosphate cotransporter 2A; Na(+)/Pi cotransporter 2A; Na+-phosphate cotransporter type II; naPi-2a; NaPi-3; renal sodium-dependent phosphate transporter; Sodium-dependent phosphate transport protein 2A; Sodium-phosphate transport protein 2A; sodium/phosphate co-transporter; Sodium/phosphate cotransporter 2A; solute carrier family 17 (sodium phosphate), member 2; solute carrier family 34 (sodium phosphate), member 1; solute carrier family 34 (type II sodium/phosphate cotransporter), member 1; Solute carrier family 34 member 1
Gene Aliases: FRTS2; HCINF2; NAPI-3; NPHLOP1; NPT2; NPTIIa; SLC11; SLC17A2; SLC34A1
UniProt ID: (Human) Q06495
Entrez Gene ID: (Human) 6569
Molecular Function:
enzyme modulator
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.