Invitrogen
Manufacturer Code:PA577010
Catalog # PIPA577010
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human SLC34A1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Na(+)-dependent phosphate cotransporter 2A; Na(+)/Pi cotransporter 2A; Na+-phosphate cotransporter type II; Na/Pi cotransporter; naPi-2a; NaPi-3; naPi-7; renal Na+/Pi transporter; renal sodium-dependent phosphate transporter; Sodium-dependent phosphate transport protein 2A; Sodium-dependent phosphate transport protein 2A Sodium-phosphate transport protein 2A Na(+)-dependent phosphate cotransporter 2A NaPi-3 Sodium/phosphate cotransporter 2A Na(+)/Pi cotransporter 2A NaPi-2a Solute carrier family 34 member 1 SLC34A1 NPT2 SLC17A2; Sodium-phosphate transport protein 2A; sodium/phosphate co-transporter; Sodium/phosphate cotransporter 2A; solute carrier family 17 (sodium phosphate), member 2; solute carrier family 17 (sodium/hydrogen exchanger) member 2; solute carrier family 17 (sodium/hydrogen exchanger), member 2; solute carrier family 34 (sodium phosphate), member 1; solute carrier family 34 (type II sodium/phosphate cotransporter), member 1; Solute carrier family 34 member 1; solute carrier family 34, member 1; type IIa Na+/Pi-cotransporter
Gene Aliases: FRTS2; HCINF2; NaPi-2; NAPI-3; NaPi-IIa; NaPi2A; NPHLOP1; NPT2; Npt2a; NPTIIa; SLC11; SLC17A2; SLC34A1
UniProt ID: (Human) Q06495, (Rat) Q06496
Entrez Gene ID: (Human) 6569, (Mouse) 20505, (Rat) 25548
Molecular Function: enzyme modulator transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.