Invitrogen
Manufacturer Code:PA5113005
Catalog # PIPA5113005
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Sodium-dependent phosphate transport protein 2A protein (1-103AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FRTS2 HCINF2 Na(+)/Pi cotransporter 2A Na(+)-dependent phosphate cotransporter 2A Na/Pi cotransporter Na+-phosphate cotransporter type II NaPi-2 NaPi2A naPi-2a NaPi-3 naPi-7 NaPi-IIa NPHLOP1 NPT2 Npt2a NPTIIa renal Na+/Pi transporter renal sodium-dependent phosphate transporter SLC11 Slc17a2 SLC34A1 sodium/phosphate co-transporter Sodium/phosphate cotransporter 2A Sodium-dependent phosphate transport protein 2A Sodium-phosphate transport protein 2A solute carrier family 17 (sodium phosphate) member 2 solute carrier family 17 (sodium/hydrogen exchanger) member 2 solute carrier family 17 (sodium/hydrogen exchanger) member 2 solute carrier family 34 (sodium phosphate) member 1 solute carrier family 34 (type II sodium/phosphate cotransporter) member 1 Solute carrier family 34 member 1 solute carrier family 34 member 1 type IIa Na+/Pi-cotransporter; Na(+)-dependent phosphate cotransporter 2A; Na(+)/Pi cotransporter 2A; Na+-phosphate cotransporter type II; Na/Pi cotransporter; naPi-2a; NaPi-3; naPi-7; renal Na+/Pi transporter; renal sodium-dependent phosphate transporter; Sodium-dependent phosphate transport protein 2A; Sodium-phosphate transport protein 2A; sodium/phosphate co-transporter; Sodium/phosphate cotransporter 2A; solute carrier family 17 (sodium phosphate), member 2; solute carrier family 17 (sodium/hydrogen exchanger), member 2; solute carrier family 34 (sodium phosphate), member 1; solute carrier family 34 (type II sodium/phosphate cotransporter), member 1; Solute carrier family 34 member 1; type IIa Na+/Pi-cotransporter
Gene Aliases: FRTS2; HCINF2; NAPI-3; NaPi-IIa; NPHLOP1; NPT2; Npt2a; NPTIIa; SLC11; SLC17A2; SLC34A1
UniProt ID: (Human) Q06495
Entrez Gene ID: (Human) 6569, (Mouse) 20505
Molecular Function: enzyme modulator transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.