Invitrogen
Manufacturer Code:PA5107264
Catalog # PIPA5107264
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide located within the following region: YVPSPQVLHR IPGTTTYAIS SLSPVALTEH SCPYGEVLEC HDPLPAKLAQ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Na(+)-dependent phosphate cotransporter 2A; Na(+)/Pi cotransporter 2A; naPi-2a; Sodium-dependent phosphate transport protein 2A; Sodium-phosphate transport protein 2A; Sodium/phosphate cotransporter 2A; solute carrier family 17 (sodium/hydrogen exchanger) member 2; solute carrier family 34 (sodium phosphate), member 1; Solute carrier family 34 member 1; solute carrier family 34, member 1
Gene Aliases: NaPi-2; NaPi2A; Npt2; Slc17a2; Slc34a1
UniProt ID: (Rat) Q06496
Entrez Gene ID: (Rat) 25548
Molecular Function:
enzyme modulator
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.