Novus Biologicals
Manufacturer Code:NBP236573UV
Catalog # NBP236573UV
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5E6.1B4 |
Immunogen | Two synthetic peptides made to internal sequences of human SLC31A1/CTR1 protein (sequences found between amino acids 50-150) [UniProt# O15431] |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: copper transport 1 homolog; Copper transporter 1; Copper transporter 1 COPT1high affinity copper uptake protein 1 CTR1MGC75487 hCTR1copper transport 1 homolog solute carrier family 31 (copper transporters) member 1 Solute carrier family 31 member 1; hCTR1; High affinity copper uptake protein 1; solute carrier family 31 (copper transporter), member 1; solute carrier family 31 (copper transporters), member 1; Solute carrier family 31 member 1
Gene Aliases: COPT1; CTR1; SLC31A1
UniProt ID: (Human) O15431
Entrez Gene ID: (Human) 1317
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.