Abnova
Manufacturer Code:PAB14558
Catalog # 89-117-142
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of human SLC31A1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Rabbit polyclonal antibody raised against synthetic peptide of SLC31A1.
For Research Use Only
Protein Aliases: copper transport 1 homolog; Copper transporter 1; COPT1; CTR1; hCTR1; High affinity copper uptake protein 1; MGC75487; solute carrier family 31 (copper transporter), member 1; solute carrier family 31 (copper transporters), member 1; Solute carrier family 31 member 1
Gene Aliases: COPT1; CTR1; SLC31A1
UniProt ID: (Human) O15431
Entrez Gene ID: (Human) 1317
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.