Proteintech
Manufacturer Code:161691AP150UL
Catalog # 50-554-319
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SLC30A8 Zinc transporter 8 ZnT 8 ZNT8; solute carrier family 30 (zinc transporter), member 8; Solute carrier family 30 member 8; Zinc transporter 8; zinc transporter ZnT-8; ZnT-8
Gene Aliases: C820002P14Rik; SLC30A8; ZnT-8; ZNT8
UniProt ID: (Human) Q8IWU4, (Rat) P0CE46, (Mouse) Q8BGG0
Entrez Gene ID: (Human) 169026, (Rat) 299903, (Mouse) 239436
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.