Novus Biologicals
Manufacturer Code:NBP29336020UL
Catalog # NB028603
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 287-376 of human SLC30A7 (NP_598003.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp686M0368 solute carrier family 30 (zinc transporter) member 7 Solute carrier family 30 member 7 zinc transporter like 2 zinc transporter ZnT-7 ZnT-7 ZNT7zinc transporter 7 ZnTL2 Znt-like transporter 2; solute carrier family 30 (zinc transporter), member 7; Solute carrier family 30 member 7; Zinc transporter 7; zinc transporter like 2; zinc transporter ZnT-7; ZnT-7; Znt-like transporter 2
Gene Aliases: SLC30A7; ZnT-7; ZNT7; ZNTL2
UniProt ID: (Human) Q8NEW0
Entrez Gene ID: (Human) 148867
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.