Invitrogen
Manufacturer Code:PA571831
Catalog # PIPA571831
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 282-310 amino acids from the Central region of human SLC30A6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: solute carrier family 30 (zinc transporter), member 4; Solute carrier family 30 member 6; Zinc transporter 6; Zinc transporter 6 ZnT-6 Solute carrier family 30 member 6 SLC30A6 ZNT6; zinc transporter-like 3; ZnT-6
Gene Aliases: 9530029F08Rik; Slc30a6; ZnT6
UniProt ID: (Mouse) Q8BJM5
Entrez Gene ID: (Mouse) 210148
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.