Abnova
Manufacturer Code:PAB16012
Catalog # 89-118-015
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus of human SLC30A6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SLC30A6.
For Research Use Only
Protein Aliases: FLJ31101; MGC45055; MST103; MSTP103; solute carrier family 30 (zinc transporter), member 6; Solute carrier family 30 member 6; Zinc transporter 6; ZnT-6; ZNT6
Gene Aliases: MST103; MSTP103; SLC30A6; ZNT6
UniProt ID: (Human) Q6NXT4
Entrez Gene ID: (Human) 55676
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.