Abnova
Manufacturer Code:H00064924M01A
Catalog # 89-107-299
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S3 |
Immunogen | SLC30A5 (AAH00808.11 a.a. &bsim 118 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant SLC30A5.
For Research Use Only
Protein Aliases: FLJ12496; FLJ12756; hZTL1; MGC5499; solute carrier family 30 (zinc transporter), member 5; Solute carrier family 30 member 5; Zinc transporter 5; zinc transporter ZTL1; ZnT-5; ZnT-like transporter 1; ZNT5; ZNTL1; ZTL1
Gene Aliases: SLC30A5; UNQ863/PRO1879; ZnT-5; ZNT5; ZNTL1; ZTL1
UniProt ID: (Human) Q8TAD4
Entrez Gene ID: (Human) 64924
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.