Invitrogen
Manufacturer Code:PA580028
Catalog # PIPA580028
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human SLC30A4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dri 27 protein; Dri 27/ZnT4 protein; Lethal milk protein; solute carrier family 30 (zinc transporter), member 4; solute carrier family 30 (zinc transporter), member 4; zinc transporter 4; Solute carrier family 30 member 4; Zinc transporter 4; Zinc transporter 4 ZnT-4 Solute carrier family 30 member 4 SLC30A4 ZNT4; ZnT-4
Gene Aliases: Lm; SLC30A4; znT-4; ZNT4
UniProt ID: (Human) O14863, (Rat) O55174, (Mouse) O35149
Entrez Gene ID: (Human) 7782, (Rat) 64469, (Mouse) 22785
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.