Novus Biologicals
Manufacturer Code:NBP294290100UL
Catalog # NB023135
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 335-429 of human SLC30A4 (NP_037441.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: solute carrier family 30 (zinc transporter) member 4 zinc transporter 4 znT-4 ZNT4Solute carrier family 30 member 4; solute carrier family 30 (zinc transporter), member 4; Solute carrier family 30 member 4; Zinc transporter 4; ZnT-4
Gene Aliases: SLC30A4; znT-4; ZNT4
UniProt ID: (Human) O14863
Entrez Gene ID: (Human) 7782
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.