Invitrogen
Manufacturer Code:PA569263
Catalog # PIPA569263
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human SLC30A3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: solute carrier family 30 (zinc transporter), member 3; Solute carrier family 30 member 3; solute carrier family 30 protein; Zinc transporter 3; zinc transporter ZnT-3; ZnT-3; ZNT3
Gene Aliases: SLC30A3; ZNT3
UniProt ID: (Bovine) Q08E25, (Human) Q99726, (Mouse) P97441, (Rat) Q6QIX3
Entrez Gene ID: (Bovine) 512803, (Human) 7781, (Horse) 100071241, (Dog) 475700, (Mouse) 22784, (Rat) 366568
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.