Novus Biologicals
Manufacturer Code:NBP297660AF750
Catalog # NB046139
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human SLC30A3 fragment. |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: solute carrier family 30 (zinc transporter) member 3 Solute carrier family 30 member 3 zinc transporter 3 znT-3 ZNT3zinc transporter ZnT-3; solute carrier family 30 (zinc transporter), member 3; Solute carrier family 30 member 3; Zinc transporter 3; zinc transporter ZnT-3; ZnT-3
Gene Aliases: SLC30A3; ZNT3
UniProt ID: (Human) Q99726
Entrez Gene ID: (Human) 7781
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.