Novus Biologicals
Manufacturer Code:NBP297795JF646
Catalog # NB047865
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human SLC30A10 fragment. |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Manganese transporter SLC30A10; member 10 solute carrier family 30 member 10 zinc transporter 10 zinc transporter 8 ZnT-10 ZNT10 ZNT8 ZRC1; Solute carrier family 30 member 10; solute carrier family 30, member 10; Zinc transporter 10; zinc transporter 8; ZnT-10
Gene Aliases: HMDPC; HMNDYT1; SLC30A10; ZnT-10; ZNT10; ZNT8; ZRC1
UniProt ID: (Human) Q6XR72
Entrez Gene ID: (Human) 55532
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.