Invitrogen
Manufacturer Code:PA577768
Catalog # PIPA577768
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)ENLEKKPRRTKAE corresponding to amino acid residues 473- 485 of rat ZnT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SLC30A1; Solute carrier family 30 member 1; solute carrier family 30, member 1; Zinc transporter 1; ZnT-1
Gene Aliases: AI839647; C130040I11Rik; Slc30a1; Znt1
UniProt ID: (Rat) Q62720, (Mouse) Q60738
Entrez Gene ID: (Rat) 58976, (Mouse) 22782
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.