Invitrogen
Manufacturer Code:PA542481
Catalog # PIPA542481
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SLC30A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Solute carrier family 30 (Zinc transporter) member 1 ZNT1 ZRC1; solute carrier family 30 (zinc transporter), member 1; Solute carrier family 30 member 1; solute carrier family 30, member 1; Zinc transporter 1; ZnT-1
Gene Aliases: SLC30A1; ZnT-1; ZNT1; ZRC1
UniProt ID: (Human) Q9Y6M5, (Rat) Q62720
Entrez Gene ID: (Pig) 503660, (Human) 7779, (Horse) 100054805, (Dog) 100286799, (Rabbit) 103350982, (Rat) 58976
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.