Abnova
Manufacturer Code:PAB12029
Catalog # 89-115-538
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 400-450 of mouse Slc2a9. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for short term. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Slc2a9.
For Research Use Only
Protein Aliases: facilitated glucose transporter, type 9; Glucose transporter type 9; GLUT-9; Glut9; Solute carrier family 2, facilitated glucose transporter member 9; Urate transporter
Gene Aliases: Glut9; Slc2a9; SLC2a9A; SLC2A9B
UniProt ID: (Mouse) Q3T9X0
Entrez Gene ID: (Mouse) 117591
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.