Invitrogen
Manufacturer Code:PA580025
Catalog # PIPA580025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of mouse Glucose Transporter 8. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glucose transporter type 8; Glucose transporter type X1; glucose transporter X1; GLUT-8; intestinal GLUT8 transporter; solute carrier family 2 (facilitated glucose transporter), member 8; Solute carrier family 2 facilitated glucose transporter member 8 Glucose transporter type 8 GLUT-8 Glucose transporter type X1 Slc2a8 Glut8 GlutX1; Solute carrier family 2, facilitated glucose transporter member 8
Gene Aliases: D2Ertd44e; GLUT8; GLUTX1; SLC2A8
UniProt ID: (Human) Q9NY64, (Mouse) Q9JIF3, (Rat) Q9JJZ1
Entrez Gene ID: (Human) 29988, (Mouse) 56017, (Rat) 85256
Molecular Function: carbohydrate transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.