Invitrogen
Manufacturer Code:PA542463
Catalog # PIPA542463
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SLC2A8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glucose transporter type 8; Glucose transporter type X1; GLUT-8; solute carrier family 2 (facilitated glucose transporter), member 8; Solute carrier family 2, facilitated glucose transporter member 8
Gene Aliases: GLUT8; GLUTX1; SLC2A8
UniProt ID: (Human) Q9NY64
Entrez Gene ID: (Human) 29988, (Rabbit) 100358726
Molecular Function:
carbohydrate transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.