Abnova
Manufacturer Code:PAB7068
Catalog # 89-132-924
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human SLC2A8. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of SLC2A8.
For Research Use Only
Protein Aliases: Glucose transporter type 8; Glucose transporter type X1; GLUT-8; GLUT8; GLUTX1; solute carrier family 2 (facilitated glucose transporter), member 8; Solute carrier family 2, facilitated glucose transporter member 8
Gene Aliases: GLUT8; GLUTX1; SLC2A8
UniProt ID: (Human) Q9NY64
Entrez Gene ID: (Human) 29988
Molecular Function:
carbohydrate transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.