Invitrogen
Manufacturer Code:PA5114399
Catalog # PIPA5114399
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human GLUT6/SLC2A6 recombinant protein (Position: M1-R507). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glucose transporter type 6; Glucose transporter type 6 glucose transporter type 9 GLUT6 GLUT-6 GLUT9 GLUT-9 HSA011372 SLC2A6 solute carrier family 2 (facilitated glucose transporter) member 6 solute carrier family 2 member 6 solute carrier family 2 facilitated glucose transporter member 6; glucose transporter type 9; GLUT-6; GLUT-9; solute carrier family 2 (facilitated glucose transporter), member 6; Solute carrier family 2, facilitated glucose transporter member 6
Gene Aliases: A330096C23; F630103L12Rik; GLUT6; GLUT9; HSA011372; SLC2A6
UniProt ID: (Human) Q5SXD7, (Mouse) Q3UDF0
Entrez Gene ID: (Human) 11182, (Rat) 296600, (Mouse) 227659
Molecular Function:
carbohydrate transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.