Invitrogen
Manufacturer Code:PA542100
Catalog # PIPA542100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SLC2A5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Fructose transporter; glucose transporter 5; Glucose transporter type 5, small intestine; glucose transporter-like protein 5; GLUT-5; GLUT5 fructose transporter; solute carrier family 2 (facilitated glucose transporter), member 5; solute carrier family 2 (facilitated glucose/fructose transporter), member 5; Solute carrier family 2, facilitated glucose transporter member 5; solute carrier family 2, member 5; testicular tissue protein Li 81
Gene Aliases: AI526984; GLUT-5; GLUT5; SLC2A5; Slc5a
UniProt ID: (Bovine) P58353, (Horse) Q863Y9, (Human) P22732, (Sheep) Q8WMN1, (Mouse) Q9WV38, (Rat) P43427
Entrez Gene ID: (Bovine) 282868, (Horse) 100033971, (Human) 6518, (Sheep) 443507, (Mouse) 56485, (Guinea pig) 100729818, (Rat) 65197
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.