Invitrogen
Manufacturer Code:PA597325
Catalog # PIPA597325
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 564-648 of human SLC2A13. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: H(+)-myo-inositol cotransporter; H(+)-myo-inositol symporter; hmit; proton (H+) myo-inositol symporter; Proton myo-inositol cotransporter; proton myo-inositol symporter; solute carrier family 2 (facilitated glucose transporter), member 13; Solute carrier family 2 member 13
Gene Aliases: 6530403A04; A630029G22Rik; AI505012; Gm308; HMIT; SLC2A13
UniProt ID: (Human) Q17S07, (Rat) Q921A2, (Mouse) Q3UHK1
Entrez Gene ID: (Human) 114134, (Rat) 171147, (Mouse) 239606
Molecular Function: carbohydrate transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.