Abnova
Manufacturer Code:PAB20351
Catalog # 89-121-851
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human SLC2A13. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant SLC2A13.
For Research Use Only
Protein Aliases: H(+)-myo-inositol cotransporter; H(+)-myo-inositol symporter; HMIT; MGC48624; proton (H+) myo-inositol symporter; Proton myo-inositol cotransporter; solute carrier family 2 (facilitated glucose transporter), member 13; Solute carrier family 2 member 13
Gene Aliases: HMIT; SLC2A13
UniProt ID: (Human) Q17S07
Entrez Gene ID: (Human) 114134
Molecular Function: carbohydrate transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.