Abnova
Manufacturer Code:PAB2256
Catalog # 89-124-269
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to amino acids 459-489 at C-terminus of human SLC29A4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SLC29A4.
For Research Use Only
Protein Aliases: ENT4; Equilibrative nucleoside transporter 4; FLJ34923; hENT4; Plasma membrane monoamine transporter; PMAT; solute carrier family 29 (equilibrative nucleoside transporter), member 4; solute carrier family 29 (nucleoside transporters), member 4; Solute carrier family 29 member 4
Gene Aliases: ENT4; PMAT; PSEC0113; SLC29A4
UniProt ID: (Human) Q7RTT9
Entrez Gene ID: (Human) 222962
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.