Invitrogen
Manufacturer Code:PA538039
Catalog # PIPA538039
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | 19 amino acid peptide near the amino terminus of human SLC29A3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Equilibrative nucleoside transporter 3; hENT3; mENT3; solute carrier family 29 (equilibrative nucleoside transporter), member 3; solute carrier family 29 (nucleoside transporters), member 3; Solute carrier family 29 member 3; Solute carrier family 29 member 3 Equilibrative nucleoside transporter 3 ENT3 HCLAP HJCD PHID
Gene Aliases: 4933435C21Rik; AW987637; ENT3; HCLAP; HJCD; PHID; SLC29A3; UNQ717/PRO1380
UniProt ID: (Human) Q9BZD2, (Mouse) Q99P65, (Rat) Q80WK7
Entrez Gene ID: (Human) 55315, (Mouse) 71279, (Rat) 353307
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.