Novus Biologicals
Manufacturer Code:NBP229615
Catalog # NBP229615
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an internal portion of the human SLC28A3 protein (between residues 30-100) [NCBI NP_071410.1] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CNT 3; CNT 3 CNT3concentrative Na+-nucleoside cotransporter Concentrative Na(+)-nucleoside cotransporter 3 hCNT3 solute carrier family 28 (sodium-coupled nucleoside transporter) member 3 solute carrier family 28 member 3; Concentrative Na(+)-nucleoside cotransporter 3; concentrative Na+-nucleoside cotransporter; solute carrier family 28 (concentrative nucleoside transporter), member 3; solute carrier family 28 (sodium-coupled nucleoside transporter), member 3; Solute carrier family 28 member 3
Gene Aliases: CNT3; SLC28A3
UniProt ID: (Human) Q9HAS3
Entrez Gene ID: (Human) 64078
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.