Invitrogen
Manufacturer Code:PA539468
Catalog # PIPA539468
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human SLC28A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CNT 2; Concentrative nucleoside transporter 2; Na(+)/nucleoside cotransporter 2; rCNT2; Sodium-coupled nucleoside transporter 2; Sodium/nucleoside cotransporter 2; Sodium/nucleoside cotransporter 2 Concentrative nucleoside transporter 2 CNT 2 hCNT2 Na(+)/nucleoside cotransporter 2 Sodium-coupled nucleoside transporter 2 Sodium/purine nucleoside co-transporter SPNT Solute carrier family 28 member 2 CNT2 CNT-2; Sodium/purine nucleoside co-transporter; Sodium/purine nucleoside cotransporter; solute carrier family 28 (concentrative nucleoside transporter), member 2; solute carrier family 28 (sodium-coupled nucleoside transporter), member 2; Solute carrier family 28 member 2; solute carrier family 28, member 2; SPNT
Gene Aliases: CNT2; HCNT2; HsT17153; SLC28A2; Spnt; SPNT1
UniProt ID: (Human) O43868, (Rat) Q62773
Entrez Gene ID: (Human) 9153, (Rat) 60423
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.