Invitrogen
Manufacturer Code:PA577546
Catalog # PIPA577546
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)ENMGAEFLESMEEGR corresponding to amino acid residues 22-36 of rat Concentrative Nucleoside Transporter 1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CNT 1; Concentrative nucleoside transporter 1; Na(+)/nucleoside cotransporter 1; Na+/nucleoside cotransporter 1; Sodium-coupled nucleoside transporter 1; Sodium/nucleoside cotransporter 1; solute carrier family 28 (sodium-coupled nucleoside transporter), member 1; Solute carrier family 28 member 1; solute carrier family 28, member 1
Gene Aliases: Cnt1; Slc28a1
UniProt ID: (Rat) Q62674
Entrez Gene ID: (Rat) 116642, (Mouse) 434203
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.