Invitrogen
Manufacturer Code:PA554023
Catalog # PIPA554023
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SLC28A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CNT 1; Concentrative nucleoside transporter 1; Na(+)/nucleoside cotransporter 1; Sodium-coupled nucleoside transporter 1; Sodium/nucleoside cotransporter 1; Sodium/nucleoside cotransporter 1 Concentrative nucleoside transporter 1 CNT 1 hCNT1 Na(+)/nucleoside cotransporter 1 Sodium-coupled nucleoside transporter 1 Solute carrier family 28 member 1 CNT1; solute carrier family 28 (concentrative nucleoside transporter), member 1; solute carrier family 28 (sodium-coupled nucleoside transporter), member 1; Solute carrier family 28 member 1
Gene Aliases: CNT1; HCNT1; SLC28A1
UniProt ID: (Human) O00337
Entrez Gene ID: (Human) 9154
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.