Novus Biologicals
Manufacturer Code:NBP230857
Catalog # NBP230857
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: MENDPSRRRESISLTPVAKGLENMGADFLESLEEGQLPRSDLSPAEIRSSWSEAAPKPFSRWRNLQPA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CNT 1; CNT 1 CNT1HCNT1 Concentrative nucleoside transporter 1 hCNT1 Na(+)/nucleoside cotransporter 1 sodium/nucleoside cotransporter 1 Sodium-coupled nucleoside transporter 1 solute carrier family 28 (sodium-coupled nucleoside transporter) member 1 Solute carrier family 28 member 1; Concentrative nucleoside transporter 1; Na(+)/nucleoside cotransporter 1; Sodium-coupled nucleoside transporter 1; Sodium/nucleoside cotransporter 1; solute carrier family 28 (concentrative nucleoside transporter), member 1; solute carrier family 28 (sodium-coupled nucleoside transporter), member 1; Solute carrier family 28 member 1
Gene Aliases: CNT1; HCNT1; SLC28A1
UniProt ID: (Human) O00337
Entrez Gene ID: (Human) 9154
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.