Invitrogen
Manufacturer Code:PA581478
Catalog # PIPA581478
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to the center region of the Mouse SLC27A3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Arachidonate--CoA ligase; FATP-3; fatty acid transport protein 3; Long-chain fatty acid transport protein 3; mFLJ00207 protein; Solute carrier family 27 member 3; Very long-chain acyl-CoA synthetase homolog 3; VLCS-3
Gene Aliases: Acsvl3; FATP3; Slc27a3; Vlcs-3
UniProt ID: (Mouse) O88561
Entrez Gene ID: (Mouse) 26568
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.