Invitrogen
Manufacturer Code:PA542490
Catalog # PIPA542490
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SLC27A3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Arachidonate--CoA ligase; FATP-3; fatty acid transport protein 3; Long-chain fatty acid transport protein 3; mFLJ00207 protein; solute carrier family 27 (fatty acid transporter), member 3; Solute carrier family 27 member 3; Very long-chain acyl-CoA synthetase homolog 3; VLCS-3
Gene Aliases: ACSVL3; FATP3; PSEC0067; SLC27A3; UNQ367/PRO703; VLCS-3
UniProt ID: (Human) Q96SW5, (Mouse) O88561
Entrez Gene ID: (Sheep) 101101860, (Dog) 100855793, (Human) 11000, (Rabbit) 100340848, (Horse) 100146983, (Guinea pig) 100726583, (Mouse) 26568, (Rat) 295219
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.