Invitrogen
Manufacturer Code:PA5110387
Catalog # PIPA5110387
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 347-646 of human SLC27A1 (NP_940982.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACSVL5 ACSVL5FATP-1 EC 6.2.1 Fatp Fatp1 FATP-1 Fatty acid transport protein FLJ00336 Long-chain fatty acid transport protein 1 MGC71751 Slc27a1 Solute carrier family 27 member 1; FATP-1; Fatty acid transport protein; fatty acid transport protein 1; Long-chain fatty acid transport protein 1; solute carrier family 27 (fatty acid transporter), member 1; Solute carrier family 27 member 1; solute carrier family 27, member 1
Gene Aliases: ACSVL5; FATP; FATP-1; FATP1; SLC27A1
UniProt ID: (Human) Q6PCB7, (Mouse) Q60714
Entrez Gene ID: (Human) 376497, (Rat) 94172, (Mouse) 26457
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.