Invitrogen
Manufacturer Code:PA5116851
Catalog # PIPA5116851
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human SLC26A9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Anion transporter/exchanger protein 9; Anion transporter/exchanger protein 9 anion transporter/exchanger-9 SLC26A9 solute carrier family 26 (anion exchanger) member 9 solute carrier family 26 member 9 solute carrier family 26 member 9; anion transporter/exchanger-9; SLC26A9 anion transporter/exchanger; solute carrier family 26 (anion exchanger), member 9; Solute carrier family 26 member 9; solute carrier family 26, member 9
Gene Aliases: E030002L01Rik; SLC26A9
UniProt ID: (Human) Q7LBE3, (Mouse) Q8BU91
Entrez Gene ID: (Human) 115019, (Mouse) 320718
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.